ENA360887303 Enamine

2-(pyridin-4-yl)propanenitrile

  • MDL number MFCD11641459
  • Cas Number : 79757-29-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98735

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98735
Availability details for
Quantity
  • Availability
  • Add to Cart