ENA360887304 Enamine

(1-benzyl-4-methylpiperidin-4-yl)methanol

  • MDL number MFCD18157617
  • Cas Number : 878167-03-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98736

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98736
Availability details for
Quantity
  • Availability
  • Add to Cart