ENA360887310 Enamine

6-chloro-2-(methylsulfanyl)pyridine-3-carboxylic acid

  • MDL number MFCD18343703
  • Cas Number : 1343727-62-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98761

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98761
Availability details for
Quantity
  • Availability
  • Add to Cart