ENA360887312 Enamine

2-(3-oxocyclohexyl)-2,3-dihydro-1H-isoindole-1,3-dione

  • MDL number MFCD19373871
  • Cas Number : 409109-10-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147...149
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96078

Safety Information

msds https://www.enaminestore.com/catalog/EN300-96078
Availability details for
Quantity
  • Availability
  • Add to Cart