ENA360887314 Enamine

4-(4-fluoro-2-nitrophenoxy)benzoic acid

  • MDL number MFCD21602499
  • Cas Number : 1304453-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96089

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96089
Availability details for
Quantity
  • Availability
  • Add to Cart