ENA360887322 Enamine

2-bromo-1-(1-methyl-1H-pyrazol-4-yl)ethan-1-one hydrobromide

  • MDL number MFCD21602404
  • Cas Number : 1252185-47-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66389

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66389
Availability details for
Quantity
  • Availability
  • Add to Cart