ENA360887324 Enamine

1-[5-chloro-2-(difluoromethoxy)phenyl]ethan-1-one

  • MDL number MFCD16661431
  • Cas Number : 1250157-06-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98795

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98795
Availability details for
Quantity
  • Availability
  • Add to Cart