ENA360887335 Enamine

1-(3-chloroprop-1-yn-1-yl)-4-(trifluoromethyl)benzene

  • MDL number MFCD12155704
  • Cas Number : 1179099-72-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98808

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98808
Availability details for
Quantity
  • Availability
  • Add to Cart