ENA360887345 Enamine

3-(1,3-thiazol-4-yl)benzonitrile

  • MDL number MFCD18275444
  • Cas Number : 1340158-15-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111...113
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98874

Safety Information

msds https://www.enaminestore.com/catalog/EN300-98874
Availability details for
Quantity
  • Availability
  • Add to Cart