ENA360887347 Enamine

2-bromo-5-(4-chlorophenyl)-1,3,4-thiadiazole

  • MDL number MFCD18344983
  • Cas Number : 57709-58-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98876

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98876
Availability details for
Quantity
  • Availability
  • Add to Cart