ENA360887354 Enamine

1-(2-bromo-5-fluorophenyl)propan-2-ol

  • MDL number MFCD16821001
  • Cas Number : 1249436-73-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98893

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98893
Availability details for
Quantity
  • Availability
  • Add to Cart