ENA360887355 Enamine

1-(3-bromophenyl)-2,2,2-trifluoroethan-1-ol

  • MDL number MFCD16067907
  • Cas Number : 446-63-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 27-29
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98894

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98894
Availability details for
Quantity
  • Availability
  • Add to Cart