ENA360887373 Enamine

1-(methylsulfanyl)propan-2-ol

  • MDL number MFCD12779642
  • Cas Number : 6943-87-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98937

Safety Information

msds https://www.enaminestore.com/catalog/EN300-98937
Availability details for
Quantity
  • Availability
  • Add to Cart