ENA360887374 Enamine

1-(5-bromothiophen-2-yl)ethan-1-amine

  • MDL number MFCD05215403
  • Cas Number : 129605-38-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98941

Safety Information

msds https://www.enaminestore.com/catalog/EN300-98941
Availability details for
Quantity
  • Availability
  • Add to Cart