ENA360887380 Enamine

methyl 4-(4-nitrophenyl)-1H-pyrrole-3-carboxylate

  • MDL number MFCD03078170
  • Cas Number : 358986-46-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-96646

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96646
Availability details for
Quantity
  • Availability
  • Add to Cart