ENA360887381 Enamine

N-hydroxycyclopropanecarboxamide

  • MDL number MFCD09937478
  • Cas Number : 5687-86-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98951

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98951
Availability details for
Quantity
  • Availability
  • Add to Cart