ENA360887382 Enamine

9H-fluorene-9-sulfonyl chloride

  • MDL number MFCD09866377
  • Cas Number : 20449-15-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98952

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98952
Availability details for
Quantity
  • Availability
  • Add to Cart