ENA360887383 Enamine

5-fluoro-2-(piperidin-4-yl)-1,3-benzothiazole

  • MDL number MFCD11636812
  • Cas Number : 1095644-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98985

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98985
Availability details for
Quantity
  • Availability
  • Add to Cart