ENA360887384 Enamine

2-hydroxyethanethioamide

  • MDL number MFCD11106303
  • Cas Number : 174971-06-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98991

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98991
Availability details for
Quantity
  • Availability
  • Add to Cart