ENA360887387 Enamine

2-(2-methylmorpholin-4-yl)-2-(thiophen-2-yl)ethan-1-amine

  • MDL number MFCD09737508
  • Cas Number : 954275-34-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71111

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71111
Availability details for
Quantity
  • Availability
  • Add to Cart