ENA360887402 Enamine

4-methylheptan-3-one

  • MDL number MFCD00039949

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99015

Safety Information

msds https://www.enaminestore.com/catalog/EN300-99015
Availability details for
Quantity
  • Availability
  • Add to Cart