ENA360887405 Enamine

4-[(dimethylamino)methyl]-3-fluoroaniline

  • MDL number MFCD20411317
  • Cas Number : 1134541-61-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-99018

Safety Information

signalword Warning
pcodes P264,P270,P280,P301+P312,P302+P352,P330,P332+P313,P362,P501
hcodes Xn
MSDS https://www.enaminestore.com/catalog/EN300-99018
pictograms GHS07
hstatements H302,H315
Availability details for
Quantity
  • Availability
  • Add to Cart