ENA360887406 Enamine

1-(2-methoxyphenyl)-3-methylbutan-2-amine

  • MDL number MFCD12782763
  • Cas Number : 1184582-52-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99019

Safety Information

msds https://www.enaminestore.com/catalog/EN300-99019
Availability details for
Quantity
  • Availability
  • Add to Cart