ENA360887418 Enamine

3-(chloromethyl)-1-methanesulfonylpiperidine

  • MDL number MFCD11612433
  • Cas Number : 1082944-42-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-99060

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99060
Availability details for
Quantity
  • Availability
  • Add to Cart