ENA360887430 Enamine

1-(2-methylpyrimidin-4-yl)piperidin-4-amine

  • MDL number MFCD17216387
  • Cas Number : 1329748-51-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99072

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99072
Availability details for
Quantity
  • Availability
  • Add to Cart