ENA360887475 Enamine

8-chloro-[1,2,3,4]tetrazolo[1,5-a]pyrazine

  • MDL number MFCD09909755
  • Cas Number : 77888-19-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96835

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96835
Availability details for
Quantity
  • Availability
  • Add to Cart