ENA360887481 Enamine

3-(carboxymethyl)-1-methyl-1H-imidazol-3-ium chloride

  • MDL number MFCD05149440
  • Cas Number : 700370-07-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 203-204
Purity 95
Salt Data Cl-
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96881

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96881
Availability details for
Quantity
  • Availability
  • Add to Cart