ENA360887497 Enamine

2-(2-phenylethyl)cyclopentan-1-one

  • MDL number MFCD01024568
  • Cas Number : 14721-44-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99160

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99160
Availability details for
Quantity
  • Availability
  • Add to Cart