ENA360887507 Enamine

[1-(aminomethyl)cyclopropyl]methanol

  • MDL number MFCD09864388
  • Cas Number : 45434-02-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81512

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-81512
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart