ENA360887512 Enamine

ethyl 2-(4-cyanophenoxy)acetate

  • MDL number MFCD01672078
  • Cas Number : 30041-95-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99209

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99209
Availability details for
Quantity
  • Availability
  • Add to Cart