ENA360887514 Enamine

2-(4-fluorophenyl)-2-(methylamino)propanoic acid

  • MDL number MFCD12047762
  • Cas Number : 1157932-41-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149...151
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99211

Safety Information

msds https://www.enaminestore.com/catalog/EN300-99211
Availability details for
Quantity
  • Availability
  • Add to Cart