ENA360887518 Enamine

2-bromo-5-(3,5-dichlorophenyl)-1,3,4-thiadiazole

  • MDL number MFCD18344977
  • Cas Number : 1339475-39-6
  • Molecular Weight : 310

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-5-(3,5-dichlorophenyl)-1,3,4-thiadiazole
Inchi Code 1S/C8H3BrCl2N2S/c9-8-13-12-7(14-8)4-1-5(10)3-6(11)2-4/h1-3H
Inchi Key MOVIEUVVVKRZHC-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-99216

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99216
Availability details for
Quantity
  • Availability
  • Add to Cart