ENA360887535 Enamine

3-ethylpiperidine hydrochloride

  • MDL number MFCD19442002
  • Cas Number : 58531-32-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-99244

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99244
Availability details for
Quantity
  • Availability
  • Add to Cart