ENA360887539 Enamine

2-[5-(propan-2-yl)-1H-1,2,3,4-tetrazol-1-yl]acetic acid

  • MDL number MFCD19282236
  • Cas Number : 1267751-57-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96933

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96933
Availability details for
Quantity
  • Availability
  • Add to Cart