ENA360887540 Enamine

2-(2-aminophenoxy)-1-(morpholin-4-yl)ethan-1-one

  • MDL number MFCD09047579
  • Cas Number : 926272-98-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111...113
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99267

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-99267
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart