ENA360887560 Enamine

4-(2-aminophenoxy)-N-methylpyridine-2-carboxamide

  • MDL number MFCD12424734
  • Cas Number : 1153328-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99295

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99295
Availability details for
Quantity
  • Availability
  • Add to Cart