ENA360887561 Enamine

5-(4-methoxyphenyl)-6-methyl-2-sulfanylpyridine-3-carbonitrile

  • MDL number MFCD19388641
  • Cas Number : 1375474-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99296

Safety Information

msds https://www.enaminestore.com/catalog/EN300-99296
Availability details for
Quantity
  • Availability
  • Add to Cart