ENA360887562 Enamine

bis(3-methoxyphenyl)methanone

  • MDL number MFCD01311539
  • Cas Number : 39193-85-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99297

Safety Information

msds https://www.enaminestore.com/catalog/EN300-99297
Availability details for
Quantity
  • Availability
  • Add to Cart