ENA360887629 Enamine

2,2-dimethyl-1-(1-methyl-1H-pyrazol-4-yl)propan-1-amine hydrochloride

  • MDL number MFCD21602814
  • Cas Number : 1375474-63-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 221...223
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99534

Safety Information

msds https://www.enaminestore.com/catalog/EN300-99534
Availability details for
Quantity
  • Availability
  • Add to Cart