ENA360887631 Enamine

methyl 5-methyl-2-oxo-2,3-dihydro-1H-imidazole-4-carboxylate

  • MDL number MFCD21362572
  • Cas Number : 53064-64-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 229...231
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99539

Safety Information

msds https://www.enaminestore.com/catalog/EN300-99539
Availability details for
Quantity
  • Availability
  • Add to Cart