ENA360887633 Enamine

1,2-dimethyl-1H,6H,7H-pyrrolo[2,3-c]pyridin-7-one

  • MDL number MFCD21602817
  • Cas Number : 1375471-39-8
  • Molecular Weight : 162.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-99555

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99555
Availability details for
Quantity
  • Availability
  • Add to Cart