ENA360887659 Enamine

1-[5-chloro-2-(difluoromethoxy)phenyl]ethan-1-ol

  • MDL number MFCD16666346
  • Cas Number : 1250521-60-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99540

Safety Information

msds https://www.enaminestore.com/catalog/EN300-99540
Availability details for
Quantity
  • Availability
  • Add to Cart