ENA360887660 Enamine

3-methylhex-5-yn-3-ol

  • MDL number MFCD18296123
  • Cas Number : 19135-01-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99542

Safety Information

msds https://www.enaminestore.com/catalog/EN300-99542
Availability details for
Quantity
  • Availability
  • Add to Cart