ENA360887661 Enamine

5-bromo-2-(4-chlorophenoxy)aniline

  • MDL number MFCD08686968
  • Cas Number : 15212-00-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99543

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99543
Availability details for
Quantity
  • Availability
  • Add to Cart