ENA360887670 Enamine

3-(furan-2-yl)propanenitrile

  • MDL number MFCD09868288
  • Cas Number : 21446-61-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99563

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99563
Availability details for
Quantity
  • Availability
  • Add to Cart