ENA360887671 Enamine

cyclopropyl(3-methoxyphenyl)methanol

  • MDL number MFCD12153715
  • Cas Number : 79134-93-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99564

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99564
Availability details for
Quantity
  • Availability
  • Add to Cart