ENA360887676 Enamine

2-(2-fluorophenoxy)butanoic acid

  • MDL number MFCD02295735
  • Cas Number : 161790-23-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63...65
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99457

Safety Information

msds https://www.enaminestore.com/catalog/EN300-99457
Availability details for
Quantity
  • Availability
  • Add to Cart