ENA360887682 Enamine

5-chloro-2-(difluoromethoxy)benzoic acid

  • MDL number MFCD06203213
  • Cas Number : 773869-44-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109...111
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99597

Safety Information

msds https://www.enaminestore.com/catalog/EN300-99597
Availability details for
Quantity
  • Availability
  • Add to Cart