ENA360887684 Enamine

4-{[(5-nitrofuran-2-yl)methyl]amino}benzoic acid

  • MDL number MFCD12714915
  • Cas Number : 88796-61-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99460

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99460
Availability details for
Quantity
  • Availability
  • Add to Cart