ENA360887686 Enamine

N-(2-hydroxypropyl)methanesulfonamide

  • MDL number MFCD11934383
  • Cas Number : 1154107-96-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99604

Safety Information

msds https://www.enaminestore.com/catalog/EN300-99604
Availability details for
Quantity
  • Availability
  • Add to Cart